missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
4-Isopropoxy-2-methyl-N-(2-pyridinyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide, Mikromol™
Beskrivelse
4-Isopropoxy-2-methyl-N-(2-pyridinyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-Isopropoxy-2-methyl-N-(2-pyridinyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide |
| CAS | 118854-49-2 |
| Analyt- eller komponentnavne | 4-Isopropoxy-2-methyl-N-(2-pyridinyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C18 H19 N3 O4 S |
| Certificeringer/compliance | ISO 17025 |
| Synonym | 4-Isopropoxy-2-methyl-N-(2-pyridinyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide |
| InChI formel | InChI=1S/C18H19N3O4S/c1-12(2)25-17-13-8-4-5-9-14(13)26(23,24)21(3)16(17)18(22)20-15-10-6-7-11-19-15/h4-12H,1-3H3,(H,19,20,22) |
| SMIL | CC(C)OC1=C(N(C)S(=O)(=O)c2ccccc12)C(=O)Nc3ccccn3 |
| IUPAC navn | 2-methyl-1,1-dioxo-4-propan-2-yloxy-N-pyridin-2-yl-1λ^{6},2-benzothiazine-3-carboxamide |
| Vis mere |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?