Få mere at vide
4-Isopropoxy-2-methyl-N-(2-pyridinyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide, Mikromol™
Artikelnummer. 30049646
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049646 100 mg
30051718 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30049646 Leverandør Mikromol Leverandørnr. MM0084.15
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-Isopropoxy-2-methyl-N-(2-pyridinyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Isopropoxy-2-methyl-N-(2-pyridinyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide
CAS 118854-49-2
Analyt- eller komponentnavne 4-Isopropoxy-2-methyl-N-(2-pyridinyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide
Forsendelsestilstand Room Temperature
Molekylær formel C18 H19 N3 O4 S
Certificeringer/compliance ISO 17025
Synonym 4-Isopropoxy-2-methyl-N-(2-pyridinyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide
InChI formel InChI=1S/C18H19N3O4S/c1-12(2)25-17-13-8-4-5-9-14(13)26(23,24)21(3)16(17)18(22)20-15-10-6-7-11-19-15/h4-12H,1-3H3,(H,19,20,22)
SMIL CC(C)OC1=C(N(C)S(=O)(=O)c2ccccc12)C(=O)Nc3ccccn3
IUPAC navn 2-methyl-1,1-dioxo-4-propan-2-yloxy-N-pyridin-2-yl-1λ^{6},2-benzothiazine-3-carboxamide
Molekylvægt (g/mol) 373.43
Mængde 100 mg
Formel vægt 373.1096 g/mol