Få mere at vide
4-Isobutyl-2-pyrrolidinone (pregabalin lactam impurity), TRC
Artikelnummer. 17914955
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17914955 50 mg
17974725 500 mg
17963445 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17914955 Leverandør Toronto Research Chemicals Leverandørnr. TRCI78650050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 61312-87-6
Molekylær formel C8 H15 N O
Molekylvægt (g/mol) 141.21
Synonym 4-(2-Methylpropyl)-2-pyrrolidinone, 4-Isobutyl-2-oxopyrrolidine, 4-Isobutyl-2-pyrrolidinone, Pregabalin-lactam
IUPAC navn 4-(2-methylpropyl)pyrrolidin-2-one
SMIL CC(C)CC1CNC(=O)C1
Tekniske data
Kemisk navn eller materiale 4-Isobutyl-2-pyrrolidinone
Renhedsgrad noter HPLC
Mængde 50 mg
InChI formel InChI=1S/C8H15NO/c1-6(2)3-7-4-8(10)9-5-7/h6-7H,3-5H2,1-2H3,(H,9,10)
Formel vægt 141.1154
Procent renhed >95%