missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Iodobenzyl bromide, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
4-Iodobenzyl bromide is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Iodobenzyl bromide is used as an organic chemical synthesis intermediate.
Solubility
Insoluble in water.
Notes
Light sensitive. Incompatible with oxidizing agents, bases and amines.
4-Iodobenzyl bromide is used as an organic chemical synthesis intermediate.
Solubility
Insoluble in water.
Notes
Light sensitive. Incompatible with oxidizing agents, bases and amines.
Specifications
Melting Point | 78°C to 82°C |
Boiling Point | 140°C (15 mmHg) |
Odor | Irritating |
Quantity | 100 g |
UN Number | UN3261 |
Beilstein | 2325160 |
Sensitivity | Light sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 296.94 |
Percent Purity | 97% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'