Learn More
4-Iodobenzyl bromide, 97%, Thermo Scientific Chemicals
Catalog Number p-7024042
Click to view available options
Quantity:
5 g
25 g
100 g
16004-15-2
C7H6BrI
296.933
MFCD00209656
BQTRMYJYYNQQGK-UHFFFAOYSA-N
4-iodobenzyl bromide, 1-bromomethyl-4-iodobenzene, 4-iodobenzylbromide, p-iodobenzyl bromide, 1-bromomethyl-4-iodo-benzene, alpha-bromo-4-iodotoluene, 4-bromomethyl-1-iodobenzene, benzene, 1-bromomethyl-4-iodo, 4-iodo benzyl bromide
954258
1-(bromomethyl)-4-iodobenzene
C1=CC(=CC=C1CBr)I
16004-15-2
C7H6BrI
296.933
MFCD00209656
BQTRMYJYYNQQGK-UHFFFAOYSA-N
4-iodobenzyl bromide, 1-bromomethyl-4-iodobenzene, 4-iodobenzylbromide, p-iodobenzyl bromide, 1-bromomethyl-4-iodo-benzene, alpha-bromo-4-iodotoluene, 4-bromomethyl-1-iodobenzene, benzene, 1-bromomethyl-4-iodo, 4-iodo benzyl bromide
954258
1-(bromomethyl)-4-iodobenzene
C1=CC(=CC=C1CBr)I
4-Iodobenzyl bromide is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Iodobenzyl bromide is used as an organic chemical synthesis intermediate.
Solubility
Insoluble in water.
Notes
Light sensitive. Incompatible with oxidizing agents, bases and amines.
TRUSTED_SUSTAINABILITY
Melting Point 78°C to 82°C
Boiling Point 140°C (15 mmHg)
Odor Irritating
Quantity 5 g
UN Number UN3261
Beilstein 2325160
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 296.94
Percent Purity 97%
Chemical Name or Material 4-Iodobenzyl bromide