Learn More
Alfa Aesar™ 4-Iodo-3-(trifluoromethyl)aniline, 98%
Brand:  Alfa Aesar™ H64426.03
Additional Details : CAS Number : 155403-06-8 Weight : 0.00100kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III
Quantity 1g
 View more versions of this product
Product Code. 15401639
  • 501.60 DKK / 1g
Estimated Shipment date
from Supplier 18-07-2018
Add to basket
Description
Description
Applications
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields and it is also an intermediate in organic syntheses.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from exposure to light. Store in dark.
Specifications
Chemical Identifiers
4-Iodo-3-(trifluoromethyl)aniline
287.02
UN2811
5-Amino-2-iodobenzotrifluoride
155403-06-8
MFCD04972659
11440140
Specifications
4-Iodo-3-(trifluoromethyl)aniline
Solid
287.02
UN2811
Off-white
5-Amino-2-iodobenzotrifluoride
155403-06-8
98%
MFCD04972659
11440140
1g
Light Sensitive