Learn More
4-Iodo-3-methyl-5-phenylisoxazole, 97%, Thermo Scientific™
Product Code. 10478182
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10478182 1 g Amber Glass Bottle 1g
10089111 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10478182 Supplier Thermo Scientific Maybridge Supplier No. CC20311DA
 Restricted Item
Forventet forsendelse: 09-12-2026
Add to basket
Chemical Identifiers
CAS 16114-53-7
Molecular Formula C10H8INO
Molecular Weight (g/mol) 285.084
MDL Number MFCD03086126
InChI Key RLZGZJOMCXKYIW-UHFFFAOYSA-N
Synonym 4-iodo-3-methyl-5-phenylisoxazole, isoxazole,4-iodo-3-methyl-5-phenyl, 3-methyl-5-phenyl-4-iodoisoxazole, 4-iodo-3-methylisoxazol-5-yl benzene, 4-iodo-3-methyl-1,2-oxazol-5-yl benzene
PubChem CID 2776520
IUPAC Name 4-iodo-3-methyl-5-phenyl-1,2-oxazole
SMILES CC1=NOC(=C1I)C2=CC=CC=C2
Specifications
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 285.08
Percent Purity 97%
Chemical Name or Material 4-Iodo-3-methyl-5-phenylisoxazole