missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-1-phenyl-1H-pyrazole, 95%, Thermo Scientific Chemicals

Catalog Number p-7031511
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
23889-85-2
C9H7IN2
270.073
MFCD11707168
PYJVDCYRDQUUKO-UHFFFAOYSA-N
4-iodo-1-phenyl-1h-pyrazole, 1-phenyl-4-iodopyrazole, 1h-pyrazole, 4-iodo-1-phenyl, pubchem13298, 4-iodanyl-1-phenyl-pyrazole, 1-phenyl-4-iodo-1h-pyrazole
14972076
4-iodo-1-phenylpyrazole
C1=CC=C(C=C1)N2C=C(C=N2)I
This item is not returnable. View return policy
23889-85-2
C9H7IN2
270.073
MFCD11707168
PYJVDCYRDQUUKO-UHFFFAOYSA-N
4-iodo-1-phenyl-1h-pyrazole, 1-phenyl-4-iodopyrazole, 1h-pyrazole, 4-iodo-1-phenyl, pubchem13298, 4-iodanyl-1-phenyl-pyrazole, 1-phenyl-4-iodo-1h-pyrazole
14972076
4-iodo-1-phenylpyrazole
C1=CC=C(C=C1)N2C=C(C=N2)I

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 270.07
Percent Purity 95%
Chemical Name or Material 4-Iodo-1-phenyl-1H-pyrazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'