missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxyquinoline-2-carboxylic acid hydrate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Broad spectrum excitatory amino acid antagonist4-Hydroxyquinoline-2-carboxylic acid hydrate is used as an intermediate in synthetic chemistry. It acts as an antiexcitotoxic and anticonvulsant. It finds application in certain neurobiological disorders therapy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in hot ethanol and water. Insoluble in ether.
Notes
Incompatible with oxidizing agents.
Soluble in hot ethanol and water. Insoluble in ether.
Notes
Incompatible with oxidizing agents.
Specifications
Melting Point | ∼275°C (decomposition) |
Quantity | 1 g |
Beilstein | 147451 |
Merck Index | 14,5327 |
Solubility Information | Soluble in hot ethanol and water. Insoluble in ether. |
IUPAC Name | 4-oxo-1H-quinoline-2-carboxylic acid;hydrate |
Formula Weight | 189.17 (Anhydrous) |
Percent Purity | 98% |
Chemical Name or Material | 4-Hydroxyquinoline-2-carboxylic acid hydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'