Få mere at vide
4-Hydroxypyridine-d5, CDN
Artikelnummer. 30319472
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319472 0.5 g
30308964 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319472 Leverandør CDN Leverandørnr. CDND51150.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Hydroxypyridine-D5
CAS 45503-33-1
Molekylær formel C5 D5 N O
Mængde 0.5 g
Synonym 4-Pyridin-2,3,5,6-d4-ol-d (9 CI), 4-Hydroxypyridine-d5, 4-Pyridinol-d5, 4-Hydroxypyridine-D5
InChI formel InChI=1 S/C5H5NO/c7-5-1-3-6-4-2-5/h1-4 H,(H,6,7)/i1D,2 D,3 D,4 D/hD
SMIL [2 H]Oc1c([2 H])c([2 H])nc([2 H])c1[2 H]
IUPAC navn 2,3,5,6-tetradeuterio-4-deuteriooxypyridine
Molekylvægt (g/mol) 100.1301
Formel vægt 100.0685 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity