Learn More
Alfa Aesar™ 4-Hydroxypyridine-2-carboxylic acid, 97%
Brand: Alfa Aesar™ H34262.06
Additional Details : CAS Number : 22468-26-4 Weight : 0.00500kg
Quantity | 5g |
---|
Description
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
slightly soluble in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Chemical Identifiers
22468-26-4 | |
139.11 | |
MXXLHBCSVDDTIX-UHFFFAOYSA-N | |
735149 | |
C1=CNC(=CC1=O)C(=O)O |
C6H5NO3 | |
MFCD00521155 | |
4-hydroxypicolinic acid, 4-hydroxypyridine-2-carboxylic acid, 4-hydroxy-2-pyridinecarboxylic acid, 4-oxo-1,4-dihydropyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 4-hydroxy, 4-hydroxypyridine-2-carboxylicacid, 2-pyridinecarboxylic acid, 1,4-dihydro-4-oxo, 4-hydroxy-picolinsaure, pubchem17131, acmc-209fwn | |
4-oxo-1H-pyridine-2-carboxylic acid |
Specifications
4-Hydroxypyridine-2-carboxylic acid | |
5g | |
C6H5NO3 | |
4-hydroxypicolinic acid, 4-hydroxypyridine-2-carboxylic acid, 4-hydroxy-2-pyridinecarboxylic acid, 4-oxo-1,4-dihydropyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 4-hydroxy, 4-hydroxypyridine-2-carboxylicacid, 2-pyridinecarboxylic acid, 1,4-dihydro-4-oxo, 4-hydroxy-picolinsaure, pubchem17131, acmc-209fwn | |
MXXLHBCSVDDTIX-UHFFFAOYSA-N | |
4-oxo-1H-pyridine-2-carboxylic acid | |
735149 | |
97% |
∽280°C (decomposition) | |
22468-26-4 | |
MFCD00521155 | |
slightly soluble in water. | |
C1=CNC(=CC1=O)C(=O)O | |
139.11 | |
139.11 |