Få mere at vide
4-Hydroxypiperidine-d9, TRC
Artikelnummer. 30407996
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30407996 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30407996 Leverandør Toronto Research Chemicals Leverandørnr. TRCH950847100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Hydroxypiperidine-d9
CAS 1219799-39-9
Molekylær formel C5H2D9NO
Mængde 100 mg
Synonym 4-Hydroxypiperidine-2,2,3,3,4,5,5,6,6-d9, 4-Piperidin-2,2,3,3,4,5,5,6,6-d9-ol (ACI)
InChI formel InChI=1S/C5H11NO/c7-5-1-3-6-4-2-5/h5-7H,1-4H2/i1D2,2D2,3D,4D2,5D/hD
SMIL OC1([2H])C([2H])([2H])C([2H])([2H])N([2H])C([2H])C1([2H])[2H]
IUPAC navn piperidin-1,2,2,3,3,4,5,5,6-d9-4-ol
Molekylvægt (g/mol) 110.2
Formel vægt 110.14