Få mere at vide
4-Hydroxypiperidine-3,3,4,5,5-d5-N-t-BOC, CDN
Artikelnummer. 30322235
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30322235 0.1 g 0.1g
30317023 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322235 Leverandør CDN Leverandørnr. CDND77930.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-(tert-Butoxycarbonyl)-4-hydroxypiperidine-3,3,4,5,5-D5
CAS 1523606-18-9
Molekylær formel C10 D5 H14 N O3
Mængde 0.1 g
Synonym 1-Piperidine-3,3,4,5,5-d5-carboxylic acid, 4-hydroxy-, 1,1-dimethylethyl ester (ACI), 1,1-Dimethylethyl 4-hydroxy-1-piperidine-3,3,4,5,5-d5-carboxylate (ACI), tert-Butyl 3,3,4,5,5-pentadeuterio-4-hydroxypiperidine-1-carboxylate, 1-(tert-Butoxycarbonyl)-4-hydroxypiperidine-3,3,4,5,5-D5, N-(tert-Butyloxycarbonyl)-4-hydroxypiperidine-3,3,4,5,5-d5, 1-t-Boc-4-hydroxypiperidine-3,3,4,5,5-d5, 4-Hydroxypiperidine-3,3,4,5,5-d5-N-t-BOC
InChI formel InChI=1 S/C10H19NO3/c1-10(2,3)14-9(13)11-6-4-8(12)5-7-11/h8,12 H,4-7H2,1-3H3/i4D2,5D2,8 D
SMIL [2 H]C1([2 H])CN(CC([2 H])([2 H])C1([2 H])O)C(=O)OC(C)(C)C
IUPAC navn tert-butyl 3,3,4,5,5-pentadeuterio-4-hydroxypiperidine-1-carboxylate
Molekylvægt (g/mol) 206.2936
Formel vægt 206.1679 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity