Få mere at vide
4-Hydroxypiperidine-3,3,4,5,5-d5, CDN
Artikelnummer. 30315789
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30315789 0.05 g -
30321290 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315789 Leverandør CDN Leverandørnr. CDND65490.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Hydroxypiperidine-3,3,4,5,5-D5
CAS 1219799-35-5
Molekylær formel C5 D5 H6 N O
Mængde 0.05 g
Synonym 3,3,4,5,5-Pentadeuteriopiperidin-4-ol, 4-Piperidin-3,3,4,5,5-d5-ol, 4-Hydroxypiperidine-3,3,4,5,5-d5, 4-Hydroxypiperidine-3,3,4,5,5-D5, 4-Piperidinol-3,3,4,5,5-d5
InChI formel InChI=1 S/C5H11NO/c7-5-1-3-6-4-2-5/h5-7 H,1-4H2/i1D2,2D2,5 D
SMIL [2 H]C1([2 H])CNCC([2 H])([2 H])C1([2 H])O
IUPAC navn 3,3,4,5,5-pentadeuteriopiperidin-4-ol
Molekylvægt (g/mol) 106.1778
Formel vægt 106.1154 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity