Få mere at vide
4’-Hydroxyphenyl Carvedilol-d3, TRC
Artikelnummer. 30404288
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30404288 10 mg
30404082 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30404288 Leverandør Toronto Research Chemicals Leverandørnr. TRCH94912110MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1189675-28-2
Molekylær formel C24 2H3 H23 N2 O5
Molekylvægt (g/mol) 425.49
Synonym 4-[2-[[3-(9H-Carbazol-4-yloxy)-2-hydroxypropyl]amino]ethoxy]-3-(trideuteriomethoxy)phenol, 4-[2-[[3-(9H-Carbazol-4-yloxy)-2-hydroxypropyl]amino]ethoxy]-3-(methoxy-d3)phenol, 4-Hydroxycarvedilol-d3 (methoxy-d3), 4'-Hydroxyphenylcarvedilol-D3 (methoxy-D3), 4'-Hydroxyphenyl carvedilol-d3
IUPAC navn 4-[2-[[3-(9H-carbazol-4-yloxy)-2-hydroxypropyl]amino]ethoxy]-3-(trideuteriomethoxy)phenol
SMIL [2H]C([2H])([2H])Oc1cc(O)ccc1OCCNCC(O)COc2cccc3[nH]c4ccccc4c23
Tekniske data
Kemisk navn eller materiale 4'-Hydroxyphenylcarvedilol-d3
Mængde 10 mg
InChI formel InChI=1S/C24H26N2O5/c1-29-23-13-16(27)9-10-21(23)30-12-11-25-14-17(28)15-31-22-8-4-7-20-24(22)18-5-2-3-6-19(18)26-20/h2-10,13,17,25-28H,11-12,14-15H2,1H3/i1D3
Formel vægt 425.203