missing translation for 'onlineSavingsMsg'
Learn More

4-(Hydroxymethyl)pyridine-2-carboxylic acid, 95%

Product Code. 15475697
Change view
Click to view available options
Mængde:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde unitSize
15475697 1 g 1g
15485697 5 g 5g
15495697 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15475697 Supplier Thermo Scientific Alfa Aesar Supplier No. H32921.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 923169-37-3 | C7H7NO3 | 153.137 g/mol

Denne Thermo Scientific Chemicals mærkevare var oprindeligt en del af Alfa Aesar produktportefølje. Noget dokumentation og etiketoplysninger kan referere til det gamle mærke. Originalen Alfa Aesar produkt-/varekode eller SKU-reference er ikke ændret som en del af mærkeovergangen til Thermo Scientific Chemicals .

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 923169-37-3
Molekylær formel C7H7NO3
Molekylvægt (g/mol) 153.137
MDL nummer MFCD08447083
InChI nøgle QMLYYPGJXCVEHA-UHFFFAOYSA-N
Synonym 4-hydroxymethyl picolinic acid, 4-hydroxymethyl pyridine-2-carboxylic acid, 4-hydroxymethyl pyridine-2-carboxylicacid
PubChem CID 22645434
IUPAC navn 4-(hydroxymethyl)pyridin-2-carboxylsyre
SMIL C1=CN=C(C=C1CO)C(=O)O

Specifications

Kemisk navn eller materiale 4-(Hydroxymethyl)pyridine-2-carboxylic acid
Smeltepunkt 178°C to 185°C (decomposition)
Mængde 1 g
Formel vægt 153.15
Procent renhed 95%

RUO– Kun brug til forskning

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.