missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxychalcone, 97%, Thermo Scientific Chemicals

Catalog Number p-7022492
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
20426-12-4
C15H12O2
224.26
MFCD00016488
PWWCDTYUYPOAIU-DHZHZOJOSA-N
4-hydroxychalcone, 4-hydroxy chalcone, 2e-3-4-hydroxyphenyl-1-phenylprop-2-en-1-one, unii-jo97q47vbu, jo97q47vbu, 3-4-hydroxyphenyl-1-phenylprop-2-en-1-one, e-3-4-hydroxyphenyl-1-phenyl-prop-2-en-1-one, 3-4-hydroxyphenyl-1-phenyl-2-propen-1-one, e-3-4-hydroxyphenyl-1-phenylprop-2-en-1-one, ccris 2228
5282361
CHEBI:34423
(E)-3-(4-hydroxyphenyl)-1-phenylprop-2-en-1-one
OC1=CC=C(\C=C\C(=O)C2=CC=CC=C2)C=C1
This item is not returnable. View return policy
20426-12-4
C15H12O2
224.26
MFCD00016488
PWWCDTYUYPOAIU-DHZHZOJOSA-N
4-hydroxychalcone, 4-hydroxy chalcone, 2e-3-4-hydroxyphenyl-1-phenylprop-2-en-1-one, unii-jo97q47vbu, jo97q47vbu, 3-4-hydroxyphenyl-1-phenylprop-2-en-1-one, e-3-4-hydroxyphenyl-1-phenyl-prop-2-en-1-one, 3-4-hydroxyphenyl-1-phenyl-2-propen-1-one, e-3-4-hydroxyphenyl-1-phenylprop-2-en-1-one, ccris 2228
5282361
CHEBI:34423
(E)-3-(4-hydroxyphenyl)-1-phenylprop-2-en-1-one
OC1=CC=C(\C=C\C(=O)C2=CC=CC=C2)C=C1

4-Hydroxychalcone is used as anti-angiogenesis, antineoplastic. It is also used as organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxychalcone is used as anti-angiogenesis, antineoplastic. It is also used as organic intermediates.

Solubility
Insoluble in water.

Notes
Store at 4°C. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 183°C to 185°C
Quantity 5 g
Beilstein 2049155
Solubility Information Insoluble in water.
Formula Weight 224.26
Percent Purity 97%
Chemical Name or Material 4-Hydroxychalcone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'