Få mere at vide
4-Hydroxybenzoic Acid-d4, TRC
Artikelnummer. 30403197
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30403197 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30403197 Leverandør Toronto Research Chemicals Leverandørnr. TRCH808452100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Hydroxybenzoic acid phenyl-d4
CAS 152404-47-2
Renhedsgrad noter HPLC
Molekylær formel C7 2H4 H2 O3
Mængde 100 mg
Synonym Benzoic-2,3,5,6-d4 acid, 4-hydroxy-, 4-Hydroxybenzoic-2,3,5,6-d4 acid, 2H4-p-Salicylic acid, 4-Hydroxybenzoic Acid-D4 (phenyl-D4), 4-Carboxyphenol-2,3,5,6-d4, 4-Hydroxybenzene-2,3,5,6-d4-carboxylic acid, p-Carboxyphenol-d4, p-Hydroxybenzoic-d4 acid, 44Hydroxy-2,3,5,6-tetradeuteriobenzoic acid
InChI formel InChI=1S/C7H6O3/c8-6-3-1-5(2-4-6)7(9)10/h1-4,8H,(H,9,10)/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c(C(=O)O)c([2H])c([2H])c1O
IUPAC navn 2,3,5,6-tetradeuterio-4-hydroxybenzoic acid
Molekylvægt (g/mol) 142.15
Formel vægt 142.0568
Procent renhed >95