Få mere at vide
4-Hydroxybenzaldehyde-alpha,2,3,5,6-d5, CDN
Artikelnummer. 30309630
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30309630 0.25 g -
30312672 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30309630 Leverandør CDN Leverandørnr. CDND74810.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Hydroxybenzaldehyde-d5
CAS 298704-22-0
Molekylær formel C7 D5 H O2
Mængde 0.25 g
Synonym Benzaldehyde-formyl,2,3,5,6-d5, 4-hydroxy- (9 CI, ACI), 4-Hydroxybenzaldehyde-formyl,2,3,5,6-d5 (ACI), 4-Hydroxybenzaldehyde-d5, 4-Hydroxybenzaldehyde-α,2,3,5,6-D5, 4-(Formyl-d)phenol-2,3,5,6-d4
InChI formel InChI=1 S/C7H6O2/c8-5-6-1-3-7(9)4-2-6/h1-5,9 H/i1D,2 D,3 D,4 D,5 D
SMIL [2 H]C(=O)c1c([2 H])c([2 H])c(O)c([2 H])c1[2 H]
IUPAC navn deuterio-(2,3,5,6-tetradeuterio-4-hydroxyphenyl)methanone
Molekylvægt (g/mol) 127.152
Formel vægt 127.068 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity