missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy Triamterene, TRC

Product Code. 30135132
Change view
Click to view available options
Mængde:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30135132 100 mg
30009454 25 mg
30005013 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135132 Supplier Toronto Research Chemicals Supplier No. TRCH971050100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1226-52-4
Molekylær formel C12 H11 N7 O
Molekylvægt (g/mol) 269.26
Synonym Phenol, 4-(2,4,7-triamino-6-pteridinyl)-, Phenol, p-(2,4,7-triamino-6-pteridinyl)- (7CI,8CI), 4-(2,4,7-Triamino-6-pteridinyl)phenol, 2,4,7-Triamino-6-(4-hydroxyphenyl)pteridine, 2,4,7-Triamino-6-(p-hydroxyphenyl)pteridine, p-Hydroxytriamterene
IUPAC navn 4-(2,4,7-triaminopteridin-6-yl)phenol
SMIL Nc1nc(N)c2nc(c(N)nc2n1)c3ccc(O)cc3

Specifikationer

Kemisk navn eller materiale 4-Hydroxy Triamterene
Mængde 100 mg
InChI formel InChI=1S/C12H11N7O/c13-9-7(5-1-3-6(20)4-2-5)16-8-10(14)18-12(15)19-11(8)17-9/h1-4,20H,(H6,13,14,15,17,18,19)
Formel vægt 269.1025
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.