Få mere at vide
4’-Hydroxy Trazodone-d6 Benzyl Ether, TRC
Artikelnummer. 30403784
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30403784 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30403784 Leverandør Toronto Research Chemicals Leverandørnr. TRCH97003210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4'-Hydroxy Trazodone-d6 Benzyl Ether
CAS 1794752-37-6
Molekylær formel C26 2H6 H22 Cl N5 O2
Mængde 10 mg
Synonym 2-[3-[4-(3-Chloro-4-benzyloxyphenyl)-1-piperazinyl]propyl-d6]-1,2,4-triazolo[4,3-a]pyridin-3(2H)-one, 2-[3-[4-(m-Chloro-p-benzyloxyphenyl)-1-piperazinyl]propyl-d6]-s-triazolo[4.3-a]pyridin-3(2H)-one, 4-benzyloxytrazodone-d6
InChI formel InChI=1S/C26H28ClN5O2/c27-23-19-22(10-11-24(23)34-20-21-7-2-1-3-8-21)30-17-15-29(16-18-30)12-6-14-32-26(33)31-13-5-4-9-25(31)28-32/h1-5,7-11,13,19H,6,12,14-18,20H2/i6D2,12D2,14D2
SMIL [2H]C([2H])(N1CCN(CC1)c2ccc(OCc3ccccc3)c(Cl)c2)C([2H])([2H])C([2H])([2H])N4N=C5C=CC=CN5C4=O
IUPAC navn 2-[3-[4-(3-chloro-4-phenylmethoxyphenyl)piperazin-1-yl]-1,1,2,2,3,3-hexadeuteriopropyl]-[1,2,4]triazolo[4,3-a]pyridin-3-one
Molekylvægt (g/mol) 484.023
Formel vægt 483.231