Få mere at vide
4-Hydroxy Toremifene, TRC
Artikelnummer. 30010798
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30010798 100 mg
30004849 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30010798 Leverandør Toronto Research Chemicals Leverandørnr. TRCH954500100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 110503-62-3
Molekylær formel C26 H28 Cl N O2
Molekylvægt (g/mol) 421.96
Synonym Phenol, 4-[(1Z)-4-chloro-1-[4-[2-(dimethylamino)ethoxy]phenyl]-2-phenyl-1-buten-1-yl]- (ACI), 4-[(1Z)-4-Chloro-1-[4-[2-(dimethylamino)ethoxy]phenyl]-2-phenyl-1-buten-1-yl]phenol (ACI), Phenol, 4-[(1Z)-4-chloro-1-[4-[2-(dimethylamino)ethoxy]phenyl]-2-phenyl-1-butenyl]- (9CI), Phenol, 4-[4-chloro-1-[4-[2-(dimethylamino)ethoxy]phenyl]-2-phenyl-1-butenyl]-, (Z)- (ZCI), (Z)-4-Hydroxytoremifene, 4-Hydroxytoremifene, 4-Hydroxy Toremifene
IUPAC navn (Z)-4-(4-chlorophenyl)-4-[4-[2-(dimethylamino)ethoxy]phenyl]-3-phenylbut-3-en-1-ol
SMIL CN(C)CCOc1ccc(cc1)\C(=C(/CCO)\c2ccccc2)\c3ccc(Cl)cc3
Tekniske data
Kemisk navn eller materiale 4-Hydroxy Toremifene
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C26H28ClNO2/c1-28(2)17-19-30-24-14-10-22(11-15-24)26(21-8-12-23(27)13-9-21)25(16-18-29)20-6-4-3-5-7-20/h3-15,29H,16-19H2,1-2H3/b26-25+
Formel vægt 421.1809
Procent renhed >95%