missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
4-Hydroxy Testosterone, TRC
Specifications
Specifications
| Kemisk navn eller materiale | 4-Hydroxytestosterone |
| CAS | 2141-17-5 |
| Molekylær formel | C19 H28 O3 |
| Mængde | 1 mg |
| Synonym | Androst-4-en-3-one, 4,17-dihydroxy-, (17β)-, Androst-4-en-3-one, 4,17β-dihydroxy- (7CI,8CI), (17β)-4,17-Dihydroxyandrost-4-en-3-one, 4-Hydroxytestosterone |
| InChI formel | InChI=1S/C19H28O3/c1-18-10-8-15(20)17(22)14(18)4-3-11-12-5-6-16(21)19(12,2)9-7-13(11)18/h11-13,16,21-22H,3-10H2,1-2H3/t11-,12-,13-,16-,18+,19-/m0/s1 |
| SMIL | C[C@]12CC[C@H]3[C@@H](CCC4=C(O)C(=O)CC[C@]34C)[C@@H]1CC[C@@H]2O |
| IUPAC navn | (8R,9S,10R,13S,14S,17S)-4,17-dihydroxy-10,13-dimethyl-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one |
| Molekylvægt (g/mol) | 304.42 |
| Formel vægt | 304.2038 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?