Få mere at vide
4'-Hydroxy Tamoxifen-d6 (contains up to 10% E isomer), TRC
Artikelnummer. 30180249
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30180249 10 mg
30179744 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30180249 Leverandør Toronto Research Chemicals Leverandørnr. TRCH95473210MG
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1346606-80-1
Molekylær formel C26H23D6NO2
Molekylvægt (g/mol) 393.55
Synonym 4-[(1Z)-1-[[4-[2-(Dimethylamino-d6)ethoxy]phenyl]phenylmethylene]propyl]phenol, 4'-Hydroxytamoxifen-d6;
SMIL OC(C=C1)=CC=C1/C(CC)=C(C2=CC=C(OCCN(C([2H])([2H])[2H])C([2H])([2H])[2H])C=C2)/C3=CC=CC=C3
Tekniske data
Kemisk navn eller materiale 4'-Hydroxy Tamoxifen-d6
Mængde 10 mg
InChI formel InChI=1S/C26H29NO2/c1-4-25(20-10-14-23(28)15-11-20)26(21-8-6-5-7-9-21)22-12-16-24(17-13-22)29-19-18-27(2)3/h5-17,28H,4,18-19H2,1-3H3/b26-25-/i2D3,3D3
Formel vægt 393.2575