Få mere at vide
4-Hydroxy Propafenone-d5 Hydrochloride, TRC
Artikelnummer. 30402940
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30402940 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30402940 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9524471MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Hydroxy Propafenone-d5 Hydrochloride
CAS 1189863-32-8
Molekylær formel C21H23D5ClNO4
Mængde 1 mg
Synonym 1-[4-Hydroxy-2-[2-hydroxy-3-(propylamino)propoxy]phenl]-3-phenyl-1-propanone-d5 Hydrochloride
InChI formel InChI=1S/C21H27NO4.ClH/c1-2-12-22-14-18(24)15-26-21-13-17(23)9-10-19(21)20(25)11-8-16-6-4-3-5-7-16;/h3-7,9-10,13,18,22-24H,2,8,11-12,14-15H2,1H3;1H/i14D2,15D2,18D;
SMIL Cl.[2H]C([2H])(NCCC)C([2H])(O)C([2H])([2H])Oc1cc(O)ccc1C(=O)CCc2ccccc2
IUPAC navn 1-[4-hydroxy-2-[1,1,2,3,3-pentadeuterio-2-hydroxy-3-(propylamino)propoxy]phenyl]-3-phenylpropan-1-one;hydrochloride
Molekylvægt (g/mol) 398.94
Formel vægt 398.2021