Få mere at vide
4'-Hydroxy-PhIP, TRC
Artikelnummer. 17940795
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17940795 1 mg
17930575 5 mg
17901395 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17940795 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9507601MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 126861-72-1
Molekylær formel C13 H12 N4 O
Molekylvægt (g/mol) 240.2606
Synonym 4-(2-Amino-1-methyl-1H-imidazo[4,5-b]pyridin-6-yl)phenol, 2-Amino-4'-hydroxy-1-methyl-6-phenylimidazo[4,5-b]pyridine
IUPAC navn 4-(2-amino-1-methylimidazo[4,5-b]pyridin-6-yl)phenol
SMIL Cn1c(N)nc2ncc(cc12)c3ccc(O)cc3
Tekniske data
Kemisk navn eller materiale 4'-Hydroxy-PhIP
Mængde 1 mg
InChI formel InChI=1S/C13H12N4O/c1-17-11-6-9(7-15-12(11)16-13(17)14)8-2-4-10(18)5-3-8/h2-7,18H,1H3,(H2,14,15,16)
Formel vægt 240.1011