Få mere at vide
4-Hydroxy Phenylbutazone-d9, TRC
Artikelnummer. 30405146
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30405146 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30405146 Leverandør Toronto Research Chemicals Leverandørnr. TRCH94006210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Hydroxy Phenylbutazone-d9
CAS 1246819-23-7
Molekylær formel C19H11D9N2O3
Mængde 10 mg
Synonym 4-Butyl-4-hydroxy-1,2-diphenyl-3,5-pyrazolidinedione-d9, 1,2-Diphenyl-4-butyl-4-hydroxypyrazolidine-3,5-dione-d9, NSC 382672-d9
InChI formel InChI=1S/C19H20N2O3/c1-2-3-14-19(24)17(22)20(15-10-6-4-7-11-15)21(18(19)23)16-12-8-5-9-13-16/h4-13,24H,2-3,14H2,1H3/i1D3,2D2,3D2,14D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C1(O)C(=O)N(N(C1=O)c2ccccc2)c3ccccc3
IUPAC navn 4-hydroxy-4-(1,1,2,2,3,3,4,4,4-nonadeuteriobutyl)-1,2-diphenylpyrazolidine-3,5-dione
Molekylvægt (g/mol) 333.43
Formel vægt 333.2039