Få mere at vide
4-Hydroxy Nisoldipine-d6, TRC
Artikelnummer. 30389801
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30389801 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30389801 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9508021MG
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Hydroxy Nisoldipine-d6
CAS 1189945-12-7
Molekylær formel C20H18D6N2O7
Mængde 1 mg
Synonym 2,6-Dimethyl-4-(2-nitro-phenyl)-1,4-dihydro-pyridine-3,5-dicarboxylic Acid 3-(2-Hydroxy-2-methylpropyl) Ester 5-Methyl Ester-d6
InChI formel InChI=1S/C20H24N2O7/c1-11-15(18(23)28-5)17(13-8-6-7-9-14(13)22(26)27)16(12(2)21-11)19(24)29-10-20(3,4)25/h6-9,17,21,25H,10H2,1-5H3/i3D3,4D3
SMIL [2H]C([2H])([2H])C(O)(COC(=O)C1=C(C)NC(=C(C1c2ccccc2[N+](=O)[O-])C(=O)OC)C)C([2H])([2H])[2H]
IUPAC navn 3-O-methyl 5-O-[3,3,3-trideuterio-2-hydroxy-2-(trideuteriomethyl)propyl] 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
Molekylvægt (g/mol) 410.45
Formel vægt 410.196