Få mere at vide
(+/-)-4-Hydroxy Mephenytoin-d3, TRC
Artikelnummer. 30407847
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30407847 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30407847 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9448772.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (+/-)-4'-Hydroxy Mephenytoin-d3
CAS 1173022-56-4
Renhedsgrad noter HPLC
Molekylær formel C12H11D3N2O3
Mængde 2.5 mg
Synonym 2,4-Imidazolidinedione, 5-ethyl-5-(4-hydroxyphenyl)-3-(methyl-d3)-, 5-Ethyl-5-(4-hydroxyphenyl)-3-(methyl-d3)-2,4-imidazolidinedione, 5-Ethyl-5-(4-hydroxyphenyl)-3-trideuteriomethyl-2,4-imidazolidinedione, (±)-4-Hydroxymephenytoin-d3, 4-Hydroxymephenytoin-d3 (methyl-d3), 4'-Hydroxymephenytoin-D3 (N-methyl-D3)
InChI formel InChI=1S/C12H14N2O3/c1-3-12(8-4-6-9(15)7-5-8)10(16)14(2)11(17)13-12/h4-7,15H,3H2,1-2H3,(H,13,17)/i2D3
SMIL [2H]C([2H])([2H])N1C(=O)NC(CC)(C1=O)c2ccc(O)cc2
IUPAC navn 5-ethyl-5-(4-hydroxyphenyl)-3-(trideuteriomethyl)imidazolidine-2,4-dione
Molekylvægt (g/mol) 237.27
Formel vægt 237.1193
Procent renhed >95