Få mere at vide
4-Hydroxy-L-(+)-2-phenylglycine, TRC
Artikelnummer. 30007582
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30007582 1 g
30006828 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30007582 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9493751G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 32462-30-9
Molekylær formel C8 H9 N O3
Molekylvægt (g/mol) 167.16
Synonym (2S)-2-Amino-2-(4-hydroxyphenyl)acetic acid, Benzeneacetic acid, α-amino-4-hydroxy-, (αS)-, Benzeneacetic acid, α-amino-4-hydroxy-, (S)-, Glycine, 2-(p-hydroxyphenyl)-, L- (8CI), (αS)-α-Amino-4-hydroxybenzeneacetic acid, (S)-(4-Hydroxyphenyl)glycine, (S)-2-(4-Hydroxyphenyl)glycine, (S)-2-Amino-2-(4-hydroxyphenyl)acetic acid, L-(+)-(p-Hydroxyphenyl)glycine, L-(+)-2-(4-Hydroxyphenyl)glycine, L-(+)-4-Hydroxyphenylglycine, L-(4-Hydroxyphenyl)glycine, L-(p-Hydroxyphenyl)glycine, L-2-(p-Hydroxyphenyl)glycine, L-α-Amino-p-hydroxyphenylacetic acid, Oxfenicine, UK 25842, p-Hydroxy-L-phenylglycine;4-Hydroxy-L-(+)-2-phenylglycine
IUPAC navn (2S)-2-amino-2-(4-hydroxyphenyl)acetic acid
SMIL N[C@H](C(=O)O)c1ccc(O)cc1
Tekniske data
Kemisk navn eller materiale (2S)-2-Amino-2-(4-hydroxyphenyl)acetic Acid
InChI formel InChI=1S/C8H9NO3/c9-7(8(11)12)5-1-3-6(10)4-2-5/h1-4,7,10H,9H2,(H,11,12)/t7-/m0/s1
Mængde 1 g
Formel vægt 167.0582