Få mere at vide
4’Hydroxy-7,8-dihydro Kavain, TRC
Artikelnummer. 30132795
Skift visning
Mængde:
2.5 mg
5 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30132795 5 mg -
30012061 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30132795 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9344505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1174388-25-0
Molekylær formel C14H16O4
Molekylvægt (g/mol) 248.27
Synonym 5,6-Dihydro-6-[2-(4-hydroxyphenyl)ethyl]-4-methoxy-2H-pyran-2-one
IUPAC navn 6-(4-hydroxyphenethyl)-4-methoxy-5,6-dihydro-2H-pyran-2-one
SMIL O=C1C=C(OC)CC(CCC2=CC=C(O)C=C2)O1
Tekniske data
Kemisk navn eller materiale 4'Hydroxy-7,8-dihydro Kavain
Mængde 5 mg
InChI formel InChI=1S/C14H16O4/c1-17-13-8-12(18-14(16)9-13)7-4-10-2-5-11(15)6-3-10/h2-3,5-6,9,12,15H,4,7-8H2,1H3
Formel vægt 248.1