missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-6-(trifluoromethyl)pyrimidine, 97%, Thermo Scientific™

Catalog Number 15401569
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1546-78-7
C5H3F3N2O
164.087
MFCD00192530
LVOYSBZJJWPUBD-UHFFFAOYSA-N
4-hydroxy-6-trifluoromethyl pyrimidine, 4-hydroxy-6-trifluoromethylpyrimidine, 6-trifluoromethyl pyrimidin-4-ol, 6-trifluoromethyl-4-pyrimidinol, 6-trifluoromethyl-1h-pyrimidin-4-one, 6-trifluoromethyl-pyrimidin-4-ol, 6-trifluoromethyl pyrimidin-4 3h-one, 6-trifluoromethyl-4-hydroxypyrimidine, 4 1h-pyrimidinone, 6-trifluoromethyl
1268071
6-(trifluoromethyl)-1H-pyrimidin-4-one
C1=C(NC=NC1=O)C(F)(F)F
This item is not returnable. View return policy
1546-78-7
C5H3F3N2O
164.087
MFCD00192530
LVOYSBZJJWPUBD-UHFFFAOYSA-N
4-hydroxy-6-trifluoromethyl pyrimidine, 4-hydroxy-6-trifluoromethylpyrimidine, 6-trifluoromethyl pyrimidin-4-ol, 6-trifluoromethyl-4-pyrimidinol, 6-trifluoromethyl-1h-pyrimidin-4-one, 6-trifluoromethyl-pyrimidin-4-ol, 6-trifluoromethyl pyrimidin-4 3h-one, 6-trifluoromethyl-4-hydroxypyrimidine, 4 1h-pyrimidinone, 6-trifluoromethyl
1268071
6-(trifluoromethyl)-1H-pyrimidin-4-one
C1=C(NC=NC1=O)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
6-(Trifluoromethyl)-4-pyrimidinol is used as building block in chemical synthesis. It may also be used in the synthesis of 10-[3-(6-trifluoromethyl-pyrimidin-4-yloxy)-propoxy]-(20S)-camptothecin.

Solubility
Solubility in methanol (almost transparency). Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat.
TRUSTED_SUSTAINABILITY
Melting Point 169°C to 171°C
Color White
Quantity 1 g
Beilstein 145548
Formula Weight 164.09
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material 4-Hydroxy-6-(trifluoromethyl)pyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'