missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-6-(trifluoromethoxy)quinoline-3-carboxylic acid, Tech., Thermo Scientific™

Catalog Number p-2701838
Click to view available options
Quantity:
1 g
10 g
50 g
This item is not returnable. View return policy
175203-86-8
C11H6F3NO4
273.167
MFCD00219848
GSUDZUNGTZIKPY-UHFFFAOYSA-N
4-hydroxy-6-trifluoromethoxy quinoline-3-carboxylic acid, 4-oxo-6-trifluoromethoxy-1h-quinoline-3-carboxylic acid, 4-hydroxy-6-trifluoromethoxy-3-quinolinecarboxylic acid, 3-quinolinecarboxylic acid, 1,4-dihydro-4-oxo-6-trifluoromethoxy, maybridge1_005008, pubchem6093, buttpark 10\01-49, 4-hydroxy-6-trifluoromethoxy quinoline-3-, 4-hydroxy-6-trifluoromethoxy-3-quinolinecarboxylicacid, 3-quinolinecarboxylicacid, 4-hydroxy-6-trifluoromethoxy
2775105
4-oxo-6-(trifluoromethoxy)-1H-quinoline-3-carboxylic acid
C1=CC2=C(C=C1OC(F)(F)F)C(=O)C(=CN2)C(=O)O
This item is not returnable. View return policy
175203-86-8
C11H6F3NO4
273.167
MFCD00219848
GSUDZUNGTZIKPY-UHFFFAOYSA-N
4-hydroxy-6-trifluoromethoxy quinoline-3-carboxylic acid, 4-oxo-6-trifluoromethoxy-1h-quinoline-3-carboxylic acid, 4-hydroxy-6-trifluoromethoxy-3-quinolinecarboxylic acid, 3-quinolinecarboxylic acid, 1,4-dihydro-4-oxo-6-trifluoromethoxy, maybridge1_005008, pubchem6093, buttpark 10\01-49, 4-hydroxy-6-trifluoromethoxy quinoline-3-, 4-hydroxy-6-trifluoromethoxy-3-quinolinecarboxylicacid, 3-quinolinecarboxylicacid, 4-hydroxy-6-trifluoromethoxy
2775105
4-oxo-6-(trifluoromethoxy)-1H-quinoline-3-carboxylic acid
C1=CC2=C(C=C1OC(F)(F)F)C(=O)C(=CN2)C(=O)O
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 273.17
Percent Purity Tech.
Grade Technical
Chemical Name or Material 4-Hydroxy-6-(trifluoromethoxy)quinoline-3-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'