missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-6-methyl-2-(trifluoromethyl)quinoline, 97%, Thermo Scientific™

Catalog Number 11449137
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1701-20-8
C11H8F3NO
227.186
MFCD00153192
UNVMZLUVACVTDT-UHFFFAOYSA-N
4-hydroxy-6-methyl-2-trifluoromethyl quinoline, 6-methyl-2-trifluoromethyl quinolin-4-ol, 6-methyl-2-trifluoromethyl-1h-quinolin-4-one, 2-trifluoromethyl-6-methylquinolin-4-ol, 6-methyl-2-trifluoromethylquinolin-4-ol, 4-quinolinol, 6-methyl-2-trifluoromethyl, 4-hydroxy-6-methyl-2-trifluoromethyl-quinoline, 6-methyl-2-trifluoromethyl-4-quinolinol, 6-methyl-2-trifluoromethyl quinolin-4 1h-one, maybridge1_004906
2775088
6-methyl-2-(trifluoromethyl)-1H-quinolin-4-one
CC1=CC2=C(C=C1)NC(=CC2=O)C(F)(F)F
This item is not returnable. View return policy
1701-20-8
C11H8F3NO
227.186
MFCD00153192
UNVMZLUVACVTDT-UHFFFAOYSA-N
4-hydroxy-6-methyl-2-trifluoromethyl quinoline, 6-methyl-2-trifluoromethyl quinolin-4-ol, 6-methyl-2-trifluoromethyl-1h-quinolin-4-one, 2-trifluoromethyl-6-methylquinolin-4-ol, 6-methyl-2-trifluoromethylquinolin-4-ol, 4-quinolinol, 6-methyl-2-trifluoromethyl, 4-hydroxy-6-methyl-2-trifluoromethyl-quinoline, 6-methyl-2-trifluoromethyl-4-quinolinol, 6-methyl-2-trifluoromethyl quinolin-4 1h-one, maybridge1_004906
2775088
6-methyl-2-(trifluoromethyl)-1H-quinolin-4-one
CC1=CC2=C(C=C1)NC(=CC2=O)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 252°C to 253°C
Quantity 1 g
Formula Weight 227.19
Percent Purity 97%
Chemical Name or Material 4-Hydroxy-6-methyl-2-(trifluoromethyl)quinoline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'