missing translation for 'onlineSavingsMsg'
Få mere at vide

4-Hydroxy-3-quinolinemethanol, TRC

Artikelnummer. 30396044
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30396044 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30396044 Leverandør Toronto Research Chemicals Leverandørnr. TRCH953160100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 4-Hydroxy-3-quinolinemethanol
CAS 82121-18-4
Molekylær formel C10H9NO2
Mængde 100 mg
Synonym 4-Hydroxy-3-(hydroxymethyl)quinoline
InChI formel InChI=1S/C10H9NO2/c12-6-7-5-11-9-4-2-1-3-8(9)10(7)13/h1-5,12H,6H2,(H,11,13)
SMIL OCC1=C(O)C2=CC=CC=C2N=C1
IUPAC navn 3-(hydroxymethyl)quinolin-4-ol
Molekylvægt (g/mol) 175.18
Formel vægt 175.06
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.