Få mere at vide
4-Hydroxy-3-nitrophenylacetic Acid-d5 (Major), TRC
Artikelnummer. 30405327
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30405327 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30405327 Leverandør Toronto Research Chemicals Leverandørnr. TRCH94868710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Hydroxy-3-nitrophenylacetic Acid-d5
CAS 929709-59-1
Molekylær formel C8H2D5NO5
Mængde 10 mg
Synonym 4-Hydroxy-3-nitrobenzeneacetic Acid-d5, 4-Hydroxy-5-nitrophenylacetic Acid-d5, Antibiotic T 0007B1-d5, Antibiotic T 000B1-d5, m-Nitro-p-hydroxyphenylacetic Acid-d5, NHPA-d5
InChI formel InChI=1S/C8H7NO5/c10-7-2-1-5(4-8(11)12)3-6(7)9(13)14/h1-3,10H,4H2,(H,11,12)/i1D,2D,3D,4D2
SMIL OC(C([2H])([2H])C1=C([2H])C([2H])=C(O)C([N+]([O-])=O)=C1[2H])=O
Molekylvægt (g/mol) 202.18
Formel vægt 202.0638