Få mere at vide
4-Hydroxy-3-methylpyridine, TRC
Artikelnummer. 30405797
Skift visning
Mængde:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30405797 5 g
30393408 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30405797 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9480755G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Hydroxy-3-methylpyridine
CAS 22280-02-0
Molekylær formel C6 H7 N O
Mængde 5 g
Synonym 4-Pyridinol, 3-methyl-, 3-Methyl-4-pyridinol, 3-Methyl-4-hydroxypyridine, 4-Hydroxy-3-methylpyridine, 3-Methylpyridin-4-ol, 4-Hydroxy-3-picoline
InChI formel InChI=1S/C6H7NO/c1-5-4-7-3-2-6(5)8/h2-4H,1H3,(H,7,8)
SMIL Cc1cnccc1O
IUPAC navn 3-methylpyridin-4-ol
Molekylvægt (g/mol) 109.126
Formel vægt 109.053