Få mere at vide
(±)-4-Hydroxy-3-methoxy-d3-mandelic Acid, CDN
Artikelnummer. 30326124
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30326124 0.05 g
30325737 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326124 Leverandør CDN Leverandørnr. CDND39190.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (±)-Vanilmandelic Acid-D3 (VMA) (4-Hydroxy-3-methoxymandelic Acid)
CAS 74495-70-8
Molekylær formel C9 2H3 H7 O5
Mængde 0.05 g
Synonym Benzeneacetic acid, α,4-dihydroxy-3-(methoxy-d3)- (9 CI), 4-Hydroxy-3-methoxy-d3-mandelic acid, 4-Hydroxy-3-methoxymandelic acid-d3, Vanillylmandelic acid-d3, VMA-D3, 4-Hydroxy-3-methoxymandelic acid D3 (methoxy-D3)
InChI formel InChI=1 S/C9H10O5/c1-14-7-4-5(2-3-6(7)10)8(11)9(12)13/h2-4,8,10-11 H,1H3,(H,12,13)/i1D3
SMIL [2 H]C([2 H])([2 H])Oc1cc(ccc1O)C(O)C(=O)O
IUPAC navn 2-hydroxy-2-[4-hydroxy-3-(trideuteriomethoxy)phenyl]acetic acid
Molekylvægt (g/mol) 201.191
Formel vægt 201.072 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity