Få mere at vide
4-Hydroxy-3-methoxy-d3-benzaldehyde O-Methyloxime, TRC
Artikelnummer. 30404440
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30404440 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30404440 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9460725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Hydroxy-3-methoxy-d3-benzaldehyde O-Methyloxime
CAS 1185238-76-9
Molekylær formel C9H8D3NO3
Mængde 5 mg
InChI formel InChI=1S/C9H11NO3/c1-12-9-5-7(6-10-13-2)3-4-8(9)11/h3-6,11H,1-2H3/i1D3
SMIL [2H]C([2H])([2H])Oc1cc(C=NOC)ccc1O
IUPAC navn 4-(methoxyiminomethyl)-2-(trideuteriomethoxy)phenol
Molekylvægt (g/mol) 184.21
Formel vægt 184.0927