Få mere at vide
4-Hydroxy-3-methoxy-4'-methyl-5-nitrobenzophenone, Mikromol™
Artikelnummer. 30460228
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30460228 4 x 25 mg
30052621 25 mg
30050815 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30460228 Leverandør Mikromol Leverandørnr. MM3329.0100254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-Hydroxy-3-methoxy-4'-methyl-5-nitrobenzophenone, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Hydroxy-3-methoxy-4'-methyl-5-nitrobenzophenone
CAS 134612-80-9
Analyt- eller komponentnavne 4-Hydroxy-3-methoxy-4'-methyl-5-nitrobenzophenone
Forsendelsestilstand Room Temperature
Molekylær formel C15 H13 N O5
Certificeringer/compliance ISO 17025
Synonym Methanone, (4-hydroxy-3-methoxy-5-nitrophenyl)(4-methylphenyl)-, 4-Hydroxy-3-methoxy-4'-methyl-5-nitrobenzophenone, Ro 40-7591, Tolcapone Related Compound B
InChI formel InChI=1S/C15H13NO5/c1-9-3-5-10(6-4-9)14(17)11-7-12(16(19)20)15(18)13(8-11)21-2/h3-8,18H,1-2H3
Løsningstype Neat
SMIL COc1cc(cc(c1O)[N+](=O)[O-])C(=O)c2ccc(C)cc2
IUPAC navn (4-hydroxy-3-methoxy-5-nitrophenyl)-(4-methylphenyl)methanone
Molekylvægt (g/mol) 287.27
Mængde 4 x 25 mg
Formel vægt 287.0794 g/mol