Få mere at vide
4-Hydroxy-3,5-bis(1-methylethyl)benzoic Acid, Mikromol™
Artikelnummer. 30050772
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050772 25 mg
30052014 100 mg
30460019 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30050772 Leverandør Mikromol Leverandørnr. MM0461.160025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-Hydroxy-3,5-bis(1-methylethyl)benzoic Acid
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Hydroxy-3,5-bis(1-methylethyl)benzoic Acid
CAS 13423-73-9
Analyt- eller komponentnavne 4-Hydroxy-3,5-bis(1-methylethyl)benzoic Acid
Forsendelsestilstand Ice pack (-20°C)
Molekylær formel C13 H18 O3
Certificeringer/compliance ISO 9001
Synonym 4-Hydroxy-3,5-bis(1-methylethyl)benzoic Acid, Benzoic acid, 4-hydroxy-3,5-diisopropyl- (6CI,8CI), 4-Hydroxy-3,5-bis(1-methylethyl)benzoic acid, 4-Hydroxy-3,5-diisopropylbenzoic acid, Propofol Imp. N (EP), Propofol Impurity N
InChI formel InChI=1S/C13H18O3/c1-7(2)10-5-9(13(15)16)6-11(8(3)4)12(10)14/h5-8,14H,1-4H3,(H,15,16)
SMIL CC(C)c1cc(cc(C(C)C)c1O)C(=O)O
IUPAC navn 4-hydroxy-3,5-di(propan-2-yl)benzoic acid
Molekylvægt (g/mol) 222.28
Mængde 25 mg
Formel vægt 222.1256 g/mol