missing translation for 'onlineSavingsMsg'
Learn More

4-[Hydroxy(2-thiazolyl)methyl]benzeneboronic acid pinacol ester, 97%, Thermo Scientific™

Catalog Number 11482150
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1073371-73-9
C16H20BNO3S
317.21
MFCD09953504
WLBDZEXNSNOBAQ-UHFFFAOYSA-N
4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl thiazol-2-yl methanol, 4-hydroxy thiazol-2-yl methyl phenylboronic acid, pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl 1,3-thiazol-2-yl methanol, 4-hydroxy 2-thiazolyl methyl benzeneboronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-thiazol-2-yl-methanol, amtb064, 4-hydroxy thiazol-2-yl methyl phenylboronic acid pinacol ester, 4-hydroxy thiazol-2-yl methyl phenylboronic acid,pinacol ester, alpha-2-thiazolyl-4-hydroxymethyl benzeneboronic acid pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl 1,3-thiazol-2-yl methanol
46739067
[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-(1,3-thiazol-2-yl)methanol
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(C3=NC=CS3)O
This item is not returnable. View return policy
1073371-73-9
C16H20BNO3S
317.21
MFCD09953504
WLBDZEXNSNOBAQ-UHFFFAOYSA-N
4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl thiazol-2-yl methanol, 4-hydroxy thiazol-2-yl methyl phenylboronic acid, pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl 1,3-thiazol-2-yl methanol, 4-hydroxy 2-thiazolyl methyl benzeneboronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-thiazol-2-yl-methanol, amtb064, 4-hydroxy thiazol-2-yl methyl phenylboronic acid pinacol ester, 4-hydroxy thiazol-2-yl methyl phenylboronic acid,pinacol ester, alpha-2-thiazolyl-4-hydroxymethyl benzeneboronic acid pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl 1,3-thiazol-2-yl methanol
46739067
[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-(1,3-thiazol-2-yl)methanol
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(C3=NC=CS3)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 147°C to 150°C
Quantity 1 g
Formula Weight 317.21
Percent Purity 97%
Chemical Name or Material 4-[Hydroxy(2-thiazolyl)methyl]benzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'