Få mere at vide
4-Hydroxy-17beta-estradiol-1,2,16,16,17-d5, CDN
Artikelnummer. 30321293
Skift visning
Mængde:
0.005 g
Artikelnummer. Mængde
30321293 0.005 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321293 Leverandør CDN Leverandørnr. CDND55490.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Hydroxy-17 beta-estradiol-16,16,17-D3
CAS 221093-38-5
Molekylær formel C18 2H5 H19 O3
Mængde 0.005 g
InChI formel InChI=1 S/C18H24O3/c1-18-9-8-11-10-4-6-15(19)17(21)13(10)3-2-12(11)14(18)5-7-16(18)20/h4,6,11-12,14,16,19-21 H,2-3,5,7-9H2,1H3/t11-,12-,14+,16+,18+/m1/s1/i4D,6 D,7D2,16 D
SMIL [2 H]c1c([2 H])c2[C@H]3 CC[C@@]4(C)[C@@H](CC([2 H])([2 H])[C@]4([2 H])O)[C@@H]3 CCc2c(O)c1O
IUPAC navn (8 R,9 S,13 S,14 S,17 S)-1,2,16,16,17-pentadeuterio-13-methyl-6,7,8,9,11,12,14,15-octahydrocyclopenta[a]phenanthrene-3,4,17-triol
Molekylvægt (g/mol) 293.41
Formel vægt 293.2039 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity