Få mere at vide
4-Hydroxy-13-cis-retinoic Acid, TRC
Artikelnummer. 30010688
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30010688 25 mg -
30009571 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30010688 Leverandør Toronto Research Chemicals Leverandørnr. TRCH95339525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 75281-25-3
Molekylær formel C20 H28 O3
Molekylvægt (g/mol) 316.43
Synonym Retinoic acid, 4-hydroxy-, 13-cis-, 13-cis-4-Hydroxyretinoic acid, 4-Hydroxy-13-cis-retinoic acid, 4-Hydroxyisotretinoin, (2Z,4E,6E,8E)-9-[(3RS)-3-Hydroxy-2,6,6-trimethylcyclohex-1-en-1-yl]-3,7-dimethylnona-2,4,6,8-tetraenoic Acid
IUPAC navn (2Z,4E,6E,8E)-9-(3-hydroxy-2,6,6-trimethylcyclohexen-1-yl)-3,7-dimethylnona-2,4,6,8-tetraenoic acid
SMIL C\C(=C\C(=O)O)\C=C\C=C(/C)\C=C\C1=C(C)C(O)CCC1(C)C
Tekniske data
Kemisk navn eller materiale 4-Hydroxy-13-cis-retinoic Acid
Mængde 25 mg
InChI formel InChI=1S/C20H28O3/c1-14(7-6-8-15(2)13-19(22)23)9-10-17-16(3)18(21)11-12-20(17,4)5/h6-10,13,18,21H,11-12H2,1-5H3,(H,22,23)/b8-6+,10-9+,14-7+,15-13-
Formel vægt 316.2038