missing translation for 'onlineSavingsMsg'
Få mere at vide

4-Hydroxy-1,3-benzenedisulfonic Acid Disodium, TRC

Artikelnummer. 30393836
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30393836 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30393836 Leverandør Toronto Research Chemicals Leverandørnr. TRCH048480100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 4-Hydroxy-1,3-benzenedisulfonic Acid Disodium
CAS 71384-45-7
Molekylær formel C6H4O7S2Na2
Mængde 100 mg
Synonym 1-Hydroxy-2,4-disulfobenzene Disodium, 4-Hydroxy-m-benzenedisulfonic Acid Disodium, NSC 227947, 1,3-Benzenedisulfonic acid, 4-hydroxy-, disodium salt (9CI), m-Benzenedisulfonic acid, 4-hydroxy-, disodium salt (7CI), Disodium 4-hydroxy-1,3-benzenedisulfonate, sodium 4-hydroxybenzene-1,3-disulfonate
InChI formel InChI=1S/C6H6O7S2.2Na/c7-5-2-1-4(14(8,9)10)3-6(5)15(11,12)13;;/h1-3,7H,(H,8,9,10)(H,11,12,13);;/q;2*+1/p-2
SMIL O=S(C1=CC(S(=O)(O[Na])=O)=C(O)C=C1)(O[Na])=O
IUPAC navn sodium 4-hydroxybenzene-1,3-disulfonate
Molekylvægt (g/mol) 298.19
Formel vægt 297.92
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.