Få mere at vide
4-Hydroperoxy Cyclophosphamide-d4, TRC
Artikelnummer. 30179935
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179935 1 mg
30179980 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30179935 Leverandør Toronto Research Chemicals Leverandørnr. TRCH7146771MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1246816-71-6
Molekylær formel C7H11D4Cl2N2O4P
Molekylvægt (g/mol) 297.11
Synonym 2-[Bis(2-chloroethyl)amino]tetrahydro-2-oxido-2H-1,3,2-oxazaphosphorin-4-yl Hydroperoxide-d4, 4-Hydroperoxycyclofosfamide-d4, Asta 6496-d4;
IUPAC navn N,N-bis(2-chloro-2,2-dideuterioethyl)-4-hydroperoxy-2-oxo-1,3,2λ^{5}-oxazaphosphinan-2-amine
SMIL O=P1(N(CC([2H])([2H])Cl)CC([2H])([2H])Cl)OCCC(OO)N1
Tekniske data
Kemisk navn eller materiale 4-Hydroperoxy Cyclophosphamide-d4
Mængde 1 mg
InChI formel InChI=1S/C7H15Cl2N2O4P/c8-2-4-11(5-3-9)16(13)10-7(15-12)1-6-14-16/h7,12H,1-6H2,(H,10,13)/i2D2,3D2
Formel vægt 296.0398