missing translation for 'onlineSavingsMsg'
Få mere at vide

4-Hydrazinobenzoic Acid-d4 Hydrochloride, TRC

Artikelnummer. 30406139
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30406139 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30406139 Leverandør Toronto Research Chemicals Leverandørnr. TRCH710152100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 4-Hydrazinobenzoic Acid-d4 Hydrochloride
CAS 1246816-67-0
Molekylær formel C7H5D4ClN2O2
Mængde 100 mg
Synonym 4-Hydrazinyl-benzoic Acid-d4 Hydrochloride, p-Hydrazino-benzoic Acid-d4 Hydrochloride, (4-Carboxyphenyl)hydrazine-d4 Monohydrochloride, 4-Carboxyphenylhydrazine-d4 Hydrochloride, 4-Carboxyphenylhydrazinium-d4 Chloride, 4-Hydrazinobenzoic Acid-d4 Hydrochloride, p-Carboxyphenylhydrazine-d4 Hydrochloride, p-Hydrazinobenzoic Acid-d4 Hydrochloride
InChI formel InChI=1S/C7H8N2O2.ClH/c8-9-6-3-1-5(2-4-6)7(10)11;/h1-4,9H,8H2,(H,10,11);1H/i1D,2D,3D,4D;
SMIL Cl.[2H]c1c([2H])c(C(=O)O)c([2H])c([2H])c1NN
IUPAC navn 2,3,5,6-tetradeuterio-4-hydrazinylbenzoic acid;hydrochloride
Molekylvægt (g/mol) 192.64
Formel vægt 192.0604
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.