missing translation for 'onlineSavingsMsg'
Learn More

4-(Furfurylaminocarbonyl)benzeneboronic acid pinacol ester, 97%, Thermo Scientific™

Catalog Number 11492270
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1073353-59-9
C18H22BNO4
327.19
MFCD09266168
ZNYVQYJVECCENX-UHFFFAOYSA-N
4-furfurylaminocarbonyl phenylboronic acid pinacol ester, n-furan-2-ylmethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 4-furfurylamino-1-carbonyl phenyl-boronic acid pinacol ester, n-furan-2-yl methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, amtb018, 4-furfurylaminocarbonyl benzeneboronic acid pinacol ester, 4-furfurylamino-1-carbonyl phenyl boronic acid pinacol ester, n-furan-2-ylmethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide
46739034
N-(furan-2-ylmethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C(=O)NCC1=CC=CO1
This item is not returnable. View return policy
1073353-59-9
C18H22BNO4
327.19
MFCD09266168
ZNYVQYJVECCENX-UHFFFAOYSA-N
4-furfurylaminocarbonyl phenylboronic acid pinacol ester, n-furan-2-ylmethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 4-furfurylamino-1-carbonyl phenyl-boronic acid pinacol ester, n-furan-2-yl methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, amtb018, 4-furfurylaminocarbonyl benzeneboronic acid pinacol ester, 4-furfurylamino-1-carbonyl phenyl boronic acid pinacol ester, n-furan-2-ylmethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide
46739034
N-(furan-2-ylmethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C(=O)NCC1=CC=CO1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 113°C to 114°C
Quantity 1 g
Formula Weight 327.18
Percent Purity 97%
Chemical Name or Material 4-(Furfurylaminocarbonyl)benzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'