Få mere at vide
4-Fluorotoluene-alpha-d1, CDN
Artikelnummer. 30317871
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30317871 0.25 g
30309050 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317871 Leverandør CDN Leverandørnr. CDND76270.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Fluorotoluene-alpha-D
CAS 131088-90-9
Molekylær formel C7 D H6 F
Mængde 0.25 g
Synonym Benzene, 1-fluoro-4-(methyl-d)- (9 CI, ACI), 1-Fluoro-4-(methyl-d)benzene (ACI), 4-Fluorotoluene-α-d1, 4-Fluorotoluene-α-D
InChI formel InChI=1 S/C7H7F/c1-6-2-4-7(8)5-3-6/h2-5 H,1H3/i1D
SMIL [2 H]Cc1ccc(F)cc1
IUPAC navn 1-(deuteriomethyl)-4-fluorobenzene
Molekylvægt (g/mol) 111.135
Formel vægt 111.0595 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity