Få mere at vide
(4-Fluorophenyl)acetic-alpha,alpha-d2 Acid, CDN
Artikelnummer. 30322171
Skift visning
Mængde:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30322171 1 g
30322078 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322171 Leverandør CDN Leverandørnr. CDND72301G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Fluorophenylacetic Acid-D2 (acetic-D2)
CAS 113715-48-3
Molekylær formel C8 D2 H5 F O2
Mængde 1 g
Synonym Benzeneacetic-d2 acid, 4-fluoro- (9 CI), (4-Fluorophenyl)acetic-α,α-d2 Acid, 4-Fluorobenzeneacetic-d2 acid, 4-Fluorophenylacetic acid-d2, 4-Fluorophenylacetic Acid-D2 (acetic-D2)
InChI formel InChI=1 S/C8H7FO2/c9-7-3-1-6(2-4-7)5-8(10)11/h1-4 H,5H2,(H,10,11)/i5D2
SMIL [2 H]C([2 H])(C(=O)O)c1ccc(F)cc1
IUPAC navn 2,2-dideuterio-2-(4-fluorophenyl)acetic acid
Molekylvægt (g/mol) 156.1507
Formel vægt 156.0556 g/mol
Procent renhed 98 atom % D, also 10% 3,5-d2), min 98% Chemical Purity